missing translation for 'onlineSavingsMsg'
Learn More

RS 67333 hydrochloride, Tocris Bioscience™

5-HT4 partial agonist

Brand:  Tocris (Bio-Techne) 098910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13428243

  • £120.54 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent and highly selective 5-HT4 partial agonist, with pKi values of 8.7 at 5-HT4 sites in guinea pig striatum and > 6 at various other receptors, including 5-HT1A, 1D, 2A, 2C, D1, D2 and M1-3. Active in vivo, with an intrinsic activity relative to 5-HT of 0.5.
Specifications

Chemical Identifiers

168986-60-5
389.361
rs 67333 hydrochloride, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hydrochloride, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butylpiperidin-4-yl propan-1-one hydrochloride, rs-67333 hydrochloride, rs 67333 hydrochloride hplc, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hcl
1-(4-amino-5-chloro-2-methoxyphenyl)-3-(1-butylpiperidin-4-yl)propan-1-one;hydrochloride
C19H30Cl2N2O2
XVBGLZNYWUUAFF-UHFFFAOYSA-N
9800491
CCCCN1CCC(CC1)CCC(=O)C2=CC(=C(C=C2OC)N)Cl.Cl

Specifications

10mg
168986-60-5
rs 67333 hydrochloride, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hydrochloride, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butylpiperidin-4-yl propan-1-one hydrochloride, rs-67333 hydrochloride, rs 67333 hydrochloride hplc, 1-4-amino-5-chloro-2-methoxyphenyl-3-1-butyl-4-piperidinyl-1-propanone hcl
CCCCN1CCC(CC1)CCC(=O)C2=CC(=C(C=C2OC)N)Cl.Cl
389.361
389.36
RS 67333 hydrochloride
C19H30Cl2N2O2
XVBGLZNYWUUAFF-UHFFFAOYSA-N
1-(4-amino-5-chloro-2-methoxyphenyl)-3-(1-butylpiperidin-4-yl)propan-1-one;hydrochloride
9800491
>99%
Videos
Documents