missing translation for 'onlineSavingsMsg'
Learn More

RS 45041-190 hydrochloride, Tocris Bioscience™

High affinity I2 ligand. Highly selective

Brand:  Tocris (Bio-Techne) 088950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458113

  • £414.72 / 50mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Highly selective I2 imidazoline receptor ligand.
Specifications

Chemical Identifiers

204274-74-8
258.146
4-chloro-2-imidazolin-2-yl isoindoline hydrochloride, 4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-1,3-dihydroisoindole hydrochloride, rs 45041-190 hydrochloride, 1h-isoindole,4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-2,3-dihydro
4-chloro-2-(4,5-dihydro-1H-imidazol-2-yl)-1,3-dihydroisoindole;hydrochloride
C11H13Cl2N3
WRYJNVYEPVATQV-UHFFFAOYSA-N
10332716
C1CN=C(N1)N2CC3=C(C2)C(=CC=C3)Cl.Cl

Specifications

RS 45041-190 hydrochloride
204274-74-8
4-chloro-2-imidazolin-2-yl isoindoline hydrochloride, 4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-1,3-dihydroisoindole hydrochloride, rs 45041-190 hydrochloride, 1h-isoindole,4-chloro-2-4,5-dihydro-1h-imidazol-2-yl-2,3-dihydro
C1CN=C(N1)N2CC3=C(C2)C(=CC=C3)Cl.Cl
258.146
258.15
50mg
C11H13Cl2N3
WRYJNVYEPVATQV-UHFFFAOYSA-N
4-chloro-2-(4,5-dihydro-1H-imidazol-2-yl)-1,3-dihydroisoindole;hydrochloride
10332716
>98%
Videos