Learn More
(RS)-3-Hydroxyphenylglycine, Tocris Bioscience™
Group I mGlu agonist
Brand:  Tocris (Bio-Techne) 032410
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13437553
  • £106.65 / 10mg
Estimated Shipment
08-07-2019
Description
Description
Agonist at PI-linked metabotropic glutamate receptors.
S-enantiomer also available.
Specifications
Chemical Identifiers
31932-87-3
167.164
amino 3-hydroxyphenyl acetic acid, 2-amino-2-3-hydroxyphenyl acetic acid, rs-3-hydroxyphenylglycine, chembl88124, rs-3h-pg, benzeneacetic acid, a-amino-3-hydroxy, 3hpg, 2-3-hydroxyphenyl glycine, amino 3-hydroxyphenyl acetic acid #, glycine, 2-m-hydroxyphenyl-, dl
2-amino-2-(3-hydroxyphenyl)acetic acid
C8H9NO3
DQLYTFPAEVJTFM-UHFFFAOYSA-N
1217
C1=CC(=CC(=C1)O)C(C(=O)O)N
Specifications
(RS)-3-Hydroxyphenylglycine
C8H9NO3
DQLYTFPAEVJTFM-UHFFFAOYSA-N
2-amino-2-(3-hydroxyphenyl)acetic acid
1217
>98%
31932-87-3
amino 3-hydroxyphenyl acetic acid, 2-amino-2-3-hydroxyphenyl acetic acid, rs-3-hydroxyphenylglycine, chembl88124, rs-3h-pg, benzeneacetic acid, a-amino-3-hydroxy, 3hpg, 2-3-hydroxyphenyl glycine, amino 3-hydroxyphenyl acetic acid #, glycine, 2-m-hydroxyphenyl-, dl
C1=CC(=CC(=C1)O)C(C(=O)O)N
167.164
167.16