missing translation for 'onlineSavingsMsg'
Learn More

RS 23597-190 hydrochloride, Tocris Bioscience™

5-HT4 antagonist

Brand:  Tocris (Bio-Techne) 072850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13427923

  • £376.32 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A high affinity, selective competitive antagonist at 5-HT4 receptors.
Specifications

Chemical Identifiers

149719-06-2
363.279
3-piperidin-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, rs 23597-190 hydrochloride, 3-piperidine-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-yl propyl4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidine-1-yl propyl-4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-azanyl-5-chloranyl-2-methoxy-benzoate hydrochloride, 4-amino-5-chloro-2-methoxy-benzoic acid 3-piperidinopropyl ester hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 3-1-piperidinyl propyl ester hydrochloride, benzoic acid, 4-amino-5-chloro-2-methoxy-, 3-piperidine-1-yl propyl ester, hydrochloride
3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate;hydrochloride
C16H24Cl2N2O3
QLZLBYYNMGQIAR-UHFFFAOYSA-N
182592
COC1=CC(=C(C=C1C(=O)OCCCN2CCCCC2)Cl)N.Cl

Specifications

50mg
149719-06-2
3-piperidin-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, rs 23597-190 hydrochloride, 3-piperidine-1-yl propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-yl propyl4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidine-1-yl propyl-4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 3-piperidin-1-ylpropyl 4-azanyl-5-chloranyl-2-methoxy-benzoate hydrochloride, 4-amino-5-chloro-2-methoxy-benzoic acid 3-piperidinopropyl ester hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 3-1-piperidinyl propyl ester hydrochloride, benzoic acid, 4-amino-5-chloro-2-methoxy-, 3-piperidine-1-yl propyl ester, hydrochloride
COC1=CC(=C(C=C1C(=O)OCCCN2CCCCC2)Cl)N.Cl
363.279
363.28
RS 23597-190 hydrochloride
C16H24Cl2N2O3
QLZLBYYNMGQIAR-UHFFFAOYSA-N
3-piperidin-1-ylpropyl 4-amino-5-chloro-2-methoxybenzoate;hydrochloride
182592
>98%
Videos
Documents