missing translation for 'onlineSavingsMsg'
Learn More

RS 17053 hydrochloride, Tocris Bioscience™

α1A antagonist

Brand:  Tocris (Bio-Techne) 098510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448233

  • £92.61 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

α1A-adrenoceptor antagonist, with very high affinity for α1A receptors (pKi and pA2 estimates of 9.1 - 9.9) and a 30 - 100 fold selectivity over the α1B and α1D subtypes (pKi and pA2 estimates 7.7 - 7.8).
Specifications

Chemical Identifiers

169505-93-5
449.416
rs 17053 hydrochloride, n-2-2-cyclopropylmethoxyphenoxy ethyl-5-chloro-?,?-dimethyl-1h-indole-3-ethanamine hydrochloride, 2-5-chloro-1h-indol-3-yl-1,1-dimethylethyl 2-2-cyclopropylmethyloxyphenoxy ethyl amine hydrochloride, 1h-indole-3-ethanamine, 5-chloro-n-2-2-cyclopropylmethoxy phenoxy ethyl-alpha,alpha-dimethyl-, monohydrochloride
1-(5-chloro-1H-indol-3-yl)-N-[2-[2-(cyclopropylmethoxy)phenoxy]ethyl]-2-methylpropan-2-amine;hydrochloride
C24H30Cl2N2O2
QFOPFGRPNPCPBX-UHFFFAOYSA-N
9824953
CC(C)(CC1=CNC2=C1C=C(C=C2)Cl)NCCOC3=CC=CC=C3OCC4CC4.Cl

Specifications

10mg
169505-93-5
rs 17053 hydrochloride, n-2-2-cyclopropylmethoxyphenoxy ethyl-5-chloro-?,?-dimethyl-1h-indole-3-ethanamine hydrochloride, 2-5-chloro-1h-indol-3-yl-1,1-dimethylethyl 2-2-cyclopropylmethyloxyphenoxy ethyl amine hydrochloride, 1h-indole-3-ethanamine, 5-chloro-n-2-2-cyclopropylmethoxy phenoxy ethyl-alpha,alpha-dimethyl-, monohydrochloride
CC(C)(CC1=CNC2=C1C=C(C=C2)Cl)NCCOC3=CC=CC=C3OCC4CC4.Cl
449.416
449.42
RS 17053 hydrochloride
C24H30Cl2N2O2
QFOPFGRPNPCPBX-UHFFFAOYSA-N
1-(5-chloro-1H-indol-3-yl)-N-[2-[2-(cyclopropylmethoxy)phenoxy]ethyl]-2-methylpropan-2-amine;hydrochloride
9824953
>98%
Videos
Documents