missing translation for 'onlineSavingsMsg'
Learn More

RS 16566 dihydrochloride, Tocris Bioscience™

5-HT3 ligand. Also shows affinity for zacopride binding site

Brand:  Tocris (Bio-Techne) 101510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488293

  • £165.00 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

High affinity ligand for the (R)-zacopride binding site (pKi 9.84 ).
Specifications

Chemical Identifiers

1217788-97-0
419.775
(R)-N-1-Azabicyclo[2.2.2]oct-3-yl-6-chloro-1-(1-methylethyl)-1H-benzimidazole-4-carboxamide
N-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-6-chloro-1-propan-2-ylbenzimidazole-4-carboxamide;dihydrochloride
C18H25Cl3N4O
LPEOWDMQSXDYRJ-CKUXDGONSA-N
90479748
CC(C)N1C=NC2=C1C=C(C=C2C(=O)NC3CN4CCC3CC4)Cl.Cl.Cl

Specifications

RS 16566 dihydrochloride
>98%
(R)-N-1-Azabicyclo[2.2.2]oct-3-yl-6-chloro-1-(1-methylethyl)-1H-benzimidazole-4-carboxamide
CC(C)N1C=NC2=C1C=C(C=C2C(=O)NC3CN4CCC3CC4)Cl.Cl.Cl
419.775
419.78
1217788-97-0
C18H25Cl3N4O
LPEOWDMQSXDYRJ-CKUXDGONSA-N
N-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]-6-chloro-1-propan-2-ylbenzimidazole-4-carboxamide;dihydrochloride
90479748
10mg
Videos