missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RS 127445 hydrochloride, Tocris Bioscience™
Selective, high affinity 5-HT2B antagonist
Brand: Tocris (Bio-Techne) 2993/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
High affinity 5-HT2B receptor antagonist (pKi = 9.5). Displays 1000-fold selectivity for 5-HT2B with good bioavailability.Chemical Identifiers
199864-86-3 | |
317.792 | |
4-4-fluoronaphthalen-1-yl-6-isopropylpyrimidin-2-amine hydrochloride, rs-127445 hydrochloride, 4-4-fluoro-1-naphthalenyl-6-1-methylethyl-2-pyrimidinamine hydrochloride, rs 127445 hydrochloride, 2-amino-4-4-fluoronaphth-1-yl-6-isopropylpyrimidine hydrochloride, mt 500 hydrochloride, rs127445 hydrochloride, rs-127445 hydrochloride hplc , powder, 2-amino-4-4-fluoronaphth-1-yl-6-isopropylpyrimidine monohydrochloride, 4-4-fluoronaphthalen-1-yl-6-isopropylpyrimidin-2-aminehydrochloride | |
4-(4-fluoronaphthalen-1-yl)-6-propan-2-ylpyrimidin-2-amine;hydrochloride |
C17H17ClFN3 | |
MKJPYBJBPRFMHL-UHFFFAOYSA-N | |
9905058 | |
CC(C)C1=NC(=NC(=C1)C2=CC=C(C3=CC=CC=C32)F)N.Cl |
Specifications
RS 127445 hydrochloride | |
199864-86-3 | |
4-4-fluoronaphthalen-1-yl-6-isopropylpyrimidin-2-amine hydrochloride, rs-127445 hydrochloride, 4-4-fluoro-1-naphthalenyl-6-1-methylethyl-2-pyrimidinamine hydrochloride, rs 127445 hydrochloride, 2-amino-4-4-fluoronaphth-1-yl-6-isopropylpyrimidine hydrochloride, mt 500 hydrochloride, rs127445 hydrochloride, rs-127445 hydrochloride hplc , powder, 2-amino-4-4-fluoronaphth-1-yl-6-isopropylpyrimidine monohydrochloride, 4-4-fluoronaphthalen-1-yl-6-isopropylpyrimidin-2-aminehydrochloride | |
CC(C)C1=NC(=NC(=C1)C2=CC=C(C3=CC=CC=C32)F)N.Cl | |
317.792 | |
317.79 |
10mg | |
C17H17ClFN3 | |
MKJPYBJBPRFMHL-UHFFFAOYSA-N | |
4-(4-fluoronaphthalen-1-yl)-6-propan-2-ylpyrimidin-2-amine;hydrochloride | |
9905058 | |
>98% |