missing translation for 'onlineSavingsMsg'
Learn More

RS 102221 hydrochloride, Tocris Bioscience™

Selective 5-HT2C antagonist

Brand:  Tocris (Bio-Techne) 1050/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13488383

  • £157.08 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Potent and selective 5-HT2C antagonist (pKi = 8.7). Displays ∽ 100-fold selectivity over the 5-HT2A and 5-HT2B subtypes and is > 100-fold selective over other 5-HT receptors, α- and β-adrenergic and muscarinic ACh receptors.
Specifications

Chemical Identifiers

187397-18-8
649.079
8-[5-(2,4-Dimethoxy-5-(4-trifluoromethylphenylsulphonamido)phenyl-5-oxopentyl]-1,3,8-triazaspiro[4.5]decane-2,4-dione hydrochloride
C27H32ClF3N4O7S
SZJZEJZLRDIHPB-UHFFFAOYSA-N
18931299

Specifications

RS 102221 hydrochloride
187397-18-8
C27H32ClF3N4O7S
SZJZEJZLRDIHPB-UHFFFAOYSA-N
N-[5-[5-(2,4-dioxo-1,3,8-triazaspiro[4.5]decan-8-yl)pentanoyl]-2,4-dimethoxyphenyl]-4-(trifluoromethyl)benzenesulfonamide;hydrochloride
18931299
10mg
>98%
8-[5-(2,4-Dimethoxy-5-(4-trifluoromethylphenylsulphonamido)phenyl-5-oxopentyl]-1,3,8-triazaspiro[4.5]decane-2,4-dione hydrochloride
COC1=CC(=C(C=C1C(=O)CCCCN2CCC3(CC2)C(=O)NC(=O)N3)NS(=O)(=O)C4=CC=C(C=C4)C(F)(F)F)OC.Cl
649.079
649.08
Videos