missing translation for 'onlineSavingsMsg'
Learn More

RS 100329 hydrochloride, Tocris Bioscience™

Potent, subtype-selective α1A antagonist

Brand:  Tocris (Bio-Techne) 132510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449153

  • £195.51 / 10mg

Estimated Shipment date
from Supplier 03-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Subtype-selective α1A-adrenoceptor antagonist (pKi = 9.6 for human cloned α1A receptors). Displays 126- and 50-fold selectivity over human α1B and α1D receptors respectively. Active in vivo.
Specifications

Chemical Identifiers

1215654-26-4
462.898
rs 100329 hydrochloride, 5-methyl-3-3-3-4-2-2,2,2-trifluoroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride, 5-methyl-3-3-3-4-2-2,2,2,-trifluroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride
5-methyl-3-[3-[4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl]propyl]-1H-pyrimidine-2,4-dione;hydrochloride
C20H26ClF3N4O3
CWVABCXVOAVUJL-UHFFFAOYSA-N
11340200
CC1=CNC(=O)N(C1=O)CCCN2CCN(CC2)C3=CC=CC=C3OCC(F)(F)F.Cl

Specifications

10mg
1215654-26-4
rs 100329 hydrochloride, 5-methyl-3-3-3-4-2-2,2,2-trifluoroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride, 5-methyl-3-3-3-4-2-2,2,2,-trifluroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride
CC1=CNC(=O)N(C1=O)CCCN2CCN(CC2)C3=CC=CC=C3OCC(F)(F)F.Cl
462.898
462.89
RS 100329 hydrochloride
C20H26ClF3N4O3
CWVABCXVOAVUJL-UHFFFAOYSA-N
5-methyl-3-[3-[4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl]propyl]-1H-pyrimidine-2,4-dione;hydrochloride
11340200
>99%
Videos
Documents