missing translation for 'onlineSavingsMsg'
Learn More

RP 001 hydrochloride, Tocris Bioscience™

Potent S1P1 agonist

Brand:  RD Systems (Bio-Techne) 4289/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13444494

  • £186.69 / 10mg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent agonist of S1P1 receptors (EC50 = 9 pM). Induces internalization and polyubiquitination of S1P1 in vitro. Selective for S1P1 over S1P2 - S1P4.
Specifications

Chemical Identifiers

1306761-53-4
468.938
rp 001 hydrochloride, n-4-5-3-cyano-4-1-methylethoxy phenyl-1,2,4-oxadiazol-3-yl-2,3-dihydro-1h-iden-1-yl-?-alanine hydrochloride
3-[[4-[5-(3-cyano-4-propan-2-yloxyphenyl)-1,2,4-oxadiazol-3-yl]-2,3-dihydro-1H-inden-1-yl]amino]propanoic acid;hydrochloride
C24H25ClN4O4
ZHPGSGYEFGEZAL-UHFFFAOYSA-N
90488927
CC(C)OC1=C(C=C(C=C1)C2=NC(=NO2)C3=C4CCC(C4=CC=C3)NCCC(=O)O)C#N.Cl

Specifications

10mg
1306761-53-4
rp 001 hydrochloride, n-4-5-3-cyano-4-1-methylethoxy phenyl-1,2,4-oxadiazol-3-yl-2,3-dihydro-1h-iden-1-yl-?-alanine hydrochloride
CC(C)OC1=C(C=C(C=C1)C2=NC(=NO2)C3=C4CCC(C4=CC=C3)NCCC(=O)O)C#N.Cl
468.938
468.93
RP 001 hydrochloride
C24H25ClN4O4
ZHPGSGYEFGEZAL-UHFFFAOYSA-N
3-[[4-[5-(3-cyano-4-propan-2-yloxyphenyl)-1,2,4-oxadiazol-3-yl]-2,3-dihydro-1H-inden-1-yl]amino]propanoic acid;hydrochloride
90488927
>98%
Videos
Documents