Learn More
Epredia™ Digital Round Tissue Section Bath
Obtain accuracy, reliability, and ease-of-use with the Epredia™ Digital Round Tissue Section Bath.
£1450.00
Specifications
Capacity (Metric) | 3.5 L |
---|---|
Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
Communications | Digital |
Description
Features
- Deep bowl for easier section pick up
- Wide bowl accommodates longer section ribbons
- Large area to rest hands when floating sections
- Miniature circuit breakers for ease-of-use; simply push to reset
- Anti-microbial paint finish (silver ion) to inhibit bacterial growth
Heating Efficiency
- Ultra-fast heating system
- Significant slide drying area at the rear of the instrument
- A secure-fitting lid reduces airborne contamination, evaporation, and improves heating efficiency
- Automatic over-temperature cut-out for safety
Specifications
3.5 L | |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
Digital |