missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Round Ice Bucket with Lid, 4L

Multi-purpose EVA buckets that provide a non-toxic alternative to conventional ice buckets

Brand:  Corning™ 432125

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15502901

  • £83.35 / Each


Estimated Shipment
28-03-2018
Add to basket

Specifications

Specifications

4L
18.4 x 26cm
1/Cs.
Orange
1
Videos