missing translation for 'onlineSavingsMsg'
Learn More

Rottlerin, Tocris Bioscience™

Reported PKCδ inhibitor

Brand:  Tocris (Bio-Techne) 161050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13429793

  • £376.32 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Originally reported to inhibit PKC isoforms. Also reported to inhibit CAM kinase III. However, recently shown to inhibit a wide range of protein kinases, and most potently to inhibit PRAK and MAPKAP-K2 (IC50 values are 1.9 and 5 μM respectively). Also shown to act as a direct mitochondrial uncoupler. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways.
Specifications

Chemical Identifiers

82-08-6
516.546
rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
CHEBI:8899
CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O
C30H28O8
DEZFNHCVIZBHBI-ZHACJKMWSA-N
5281847
(E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one

Specifications

Rottlerin
82-08-6
rottlerin, mallotoxin, kamalin, unii-e29lp3zmuh, e29lp3zmuh, chembl34241, bmk1-f10, e-1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one, 1-6-3-acetyl-2,4,6-trihydroxy-5-methyl-benzyl-5,7-dihydroxy-2,2-dimethyl-2h-chromen-8-yl-3-phenyl-propenone, 1-6-3-acetyl-2,4,6-trihydroxy-5-methylphenyl methyl-5,7-dihydroxy-2,2-dimethyl-2h-1-benzopyran-8-yl-3-phenyl-2-propen-1-one
CC1=C(C(=C(C(=C1O)C(=O)C)O)CC2=C(C3=C(C(=C2O)C(=O)C=CC4=CC=CC=C4)OC(C=C3)(C)C)O)O
516.546
CHEBI:8899
>98%
50mg
C30H28O8
DEZFNHCVIZBHBI-ZHACJKMWSA-N
(E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethylchromen-8-yl]-3-phenylprop-2-en-1-one
5281847
516.55
Videos
Documents