missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor S-4xUniversal
Rotor S-4xUniversal is a high-capacity, swing bucket rotor suitable for use with the 5910 R refrigerated centrifuge.
£1555.00 - £3490.00
Specifications
Capacity | 4 to 750 mL |
---|---|
Max. Speed | 4500 rpm |
For Use With (Equipment) | 5910 R Centrifuge |
No. of Places | 4 |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15873212
|
Eppendorf™
5895201008 |
Without buckets |
£1555.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15863212
|
Eppendorf™
5895200001 |
Universal Buckets |
£3490.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- High capacity swing-bucket rotor
- Broad range of adapters enables high versatility for tubes, flasks, and plates
- Universal bucket design accommodates both plates and tubes
- Universal adapters accommodate selective plate formats and conical tubes
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
- Requires buckets and adapters to accommodate tubes and flasks from 4 to 750 mL and plates
Specifications
4 to 750 mL | |
4500 rpm | |
5910 R Centrifuge | |
4 |