missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rotor Packages for Thermo Scientific™ Sorvall™ ST8 Benchtop Centrifuge

Convenient cell culture and clinical packages

Brand:  Thermo Scientific™ 75005761

Code : NEW

Additional Details : Weight : 4.53590kg

 View more versions of this product

Product Code. 15145133

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Aluminum
8 x 50mL
4,500rpm
12.4cm
Thermo Scientific™ Sorvall™ ST8 benchtop Centrifuge
TX-150 Rotor (75005701), Set of 4 Conical Buckets (75005703), Set of 2 Adapters for 15mL Conical Tubes (75005808)
Swinging Bucket Rotor
3,260 x g
5.1 to 14.4cm
1.8cm
90°
Videos
View accessory products