missing translation for 'onlineSavingsMsg'
Learn More

Buchi Rotary evaporator with heating bath 230V 50Hz Rotavapor R-250EX, double reflux assembly

Brand:  Buchi 8621C511

Rotary evaporator with heating bath 230V 50Hz Rotavapor R-250EX, double reflux assembly

Code : 62

Additional Details : Weight : 250.00000kg

Product Code. 12636849

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.