missing translation for 'onlineSavingsMsg'
Learn More

Buchi Rotary evaporator with heating bath 230V 50Hz Rotavapor R-250EX, double reflux Bullfrog assembly

Brand:  Buchi 8621C521

Rotary evaporator with heating bath 230V 50Hz Rotavapor R-250EX, double reflux Bullfrog assembly

Code : 62

Additional Details : Weight : 250.00000kg

Product Code. 12656849

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.