missing translation for 'onlineSavingsMsg'
Learn More

ROS 234 dioxalate, Tocris Bioscience™

Potent H3 antagonist

Brand:  Tocris (Bio-Techne) 203410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459393

  • £113.40 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent H3 antagonist (pKB at guinea-pig ileum H3-receptor = 9.46). Limited blood brain barrier permeability (ED50 = 19.12 mg/kg i.p.).
Specifications

Chemical Identifiers

184576-87-2
421.366
ros 234 dioxalate, n-3-1h-imidazol-4-yl propyl-1h-benzimidazol-2-amine dioxalate
N-[3-(1H-imidazol-5-yl)propyl]-1H-benzimidazol-2-amine;oxalic acid
C17H19N5O8
JKTBCMILCFDKEJ-UHFFFAOYSA-N
56972188
C1=CC=C2C(=C1)NC(=N2)NCCCC3=CN=CN3.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O

Specifications

ROS 234 dioxalate
184576-87-2
ros 234 dioxalate, n-3-1h-imidazol-4-yl propyl-1h-benzimidazol-2-amine dioxalate
C1=CC=C2C(=C1)NC(=N2)NCCCC3=CN=CN3.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O
421.366
421.37
10mg
C17H19N5O8
JKTBCMILCFDKEJ-UHFFFAOYSA-N
N-[3-(1H-imidazol-5-yl)propyl]-1H-benzimidazol-2-amine;oxalic acid
56972188
>99%
Videos