Learn More
ROR gamma (t) Monoclonal Antibody (B2D), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£167.00 - £335.00
Specifications
Antigen | ROR gamma (t) |
---|---|
Clone | B2D |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15391380
|
Affymetrix eBioscience
46-6981-80 |
25 μg |
£167.00
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15301390
|
Affymetrix eBioscience
46-6981-82 |
100 μg |
£335.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The B2D monoclonal antibody reacts with the mouse ROR gamma (t) protein. RORgamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes ROR alpha and ROR beta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. ROR gamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, ROR gamma t, which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ double-positive thymoyctes, Th17 cells of the periphery and lymphoid tissue inducer (Lti) cells of lymphoid organs. The ROR gamma t isoform differs from RORgamma by three unique amino acids at the amino terminus. Therefore, the B2D antibody will react with both the ROR gamma and ROR gamma t isoforms. Applications Reported: This B2D antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This B2D antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
ROR gamma (t) Monoclonal antibody specifically detects ROR gamma (t) in Mouse samples. It is validated for Flow Cytometry.Specifications
ROR gamma (t) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
EGK_01272; IMD42; MGC129539; NR1F3; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; Rorc; Rorg; RORgamma; RORgt; RP11-98D18.11-001; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR | |
Rorc | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Rorc |
B2D | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
P51450 | |
19885 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.