Learn More
ROR gamma (t) Monoclonal Antibody (B2D), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 61-6981-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The B2D monoclonal antibody reacts with the mouse ROR gamma (t) protein. ROR gamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes ROR alpha and ROR beta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. ROR gamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, ROR gamma t, which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ "double-positive" thymoyctes, Th17 cells of the periphery and lymphoid tissue inducer (Lti) cells of lymphoid organs. The ROR gamma t isoform differs from ROR gamma by three unique amino acids at the amino terminus. Therefore, the B2D antibody will react with both the ROR gamma and ROR gamma t isoforms. Applications Reported: This B2D antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This B2D antibody has been tested by intracellular staining and flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular protein under the Resources Tab online. This can be used at less than or equal to 0.25 μg per test.
ROR gamma (t) Monoclonal antibody specifically detects ROR gamma (t) in Mouse samples. It is validated for Flow Cytometry.Specifications
ROR gamma (t) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P51450 | |
Rorc | |
Affinity chromatography | |
RUO | |
19885 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
B2D | |
PE-eFluor 610 | |
Rorc | |
EGK_01272; IMD42; MGC129539; NR1F3; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; Rorc; Rorg; RORgamma; RORgt; RP11-98D18.11-001; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.