missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
POA
Specifications
Antigen | ROBO2 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ROBO2 Ser22-Ala312, RPVA, Pro313-Pro859 Accession # Q9HCK4 |
Gene ID (Entrez) | 6092 |
Gene Accession No. | Q9HCK4 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
ROBO2 Polyclonal specifically detects ROBO2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
ROBO2 | |
6092 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human ROBO2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO3 and rhROBO4 is observed. | |
Unconjugated | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human ROBO2 Ser22-Ala312, RPVA, Pro313-Pro859 Accession # Q9HCK4 | |
Q9HCK4 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only