missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robenidine hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 252.0°C to 254.0°C |
Quantity | 1 g |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Assay Percent Range | 97.5% min. (HPLC) |
Formula Weight | 370.66 |
Percent Purity | 98% |
Chemical Name or Material | Robenidine hydrochloride |
Safety and Handling
Product Identifier
- Robenidine hydrochloride
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'