missing translation for 'onlineSavingsMsg'
Learn More

Ro 90-7501, Tocris Bioscience™

Inhibitor of Aβ42 fibril formation

Brand:  Tocris (Bio-Techne) 240850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13441154

  • £654.16 / 50mg

Estimated Shipment
11-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of amyloid β42 (Aβ42) fibril assembly; reduces Aβ42-induced toxicity (EC50 = 2 μM).
Specifications

Chemical Identifiers

293762-45-5
340.39
gnf-pf-5510, 2'-4-amino-phenyl-3h,3'h-2,5' bibenzoimidazolyl-5-ylamine, 2-2-4-aminophenyl-3h-1,3-benzodiazol-5-yl-3h-1,3-benzodiazol-5-amine, cbchromo1_000149, cambridge id 5180073, ro hplc, 2'-4-aminophenyl-2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl-1h,1'h-2,5'-bibenzimidazol-5-amine, 2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl, 2-2-4-aminophenyl benzimidazol-6-yl benzimidazole-6-ylamine
2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine
C20H16N6
PAGZCEHLFCJSPV-UHFFFAOYSA-N
824226
C1=CC(=CC=C1C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N)N

Specifications

Ro 90-7501
293762-45-5
gnf-pf-5510, 2'-4-amino-phenyl-3h,3'h-2,5' bibenzoimidazolyl-5-ylamine, 2-2-4-aminophenyl-3h-1,3-benzodiazol-5-yl-3h-1,3-benzodiazol-5-amine, cbchromo1_000149, cambridge id 5180073, ro hplc, 2'-4-aminophenyl-2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl-1h,1'h-2,5'-bibenzimidazol-5-amine, 2,5'-bi-1h-benzimidazol-5-amine, 2'-4-aminophenyl, 2-2-4-aminophenyl benzimidazol-6-yl benzimidazole-6-ylamine
C1=CC(=CC=C1C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N)N
340.39
340.39
50mg
C20H16N6
PAGZCEHLFCJSPV-UHFFFAOYSA-N
2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine
824226
>99%
Videos
Documents