missing translation for 'onlineSavingsMsg'
Learn More

Ro 67-7476, Tocris Bioscience™

Positive allosteric modulator of mGlu1

Brand:  RD Systems (Bio-Techne) 4346/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464594

  • £652.77 / 50mg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Positive allosteric modulator of mGlu 1 receptors. Potentiates glutamate-induced calcium release (EC 50 = 60.1 nM at rat mGluR1a). Displays no activity at human mGlu 1 receptors.
Specifications

Chemical Identifiers

298690-60-5
319.394
s-2-4-fluoro-phenyl-1-toluene-4-sulfonyl-pyrrolidine, pyrrolidine, 2-4-fluorophenyl-1-4-methylphenyl sulfonyl-, 2s, 2s-2-4-fluorophenyl-1-4-methylbenzenesulfonyl pyrrolidine, 2s-2-4-fluorophenyl-1-4-methylbenzene-1-sulfonyl pyrrolidine, 2s-2-4-fluorophenyl-1-4-methylphenyl sulfonyl-pyrrolidine
(2S)-2-(4-fluorophenyl)-1-(4-methylphenyl)sulfonylpyrrolidine
C17H18FNO2S
DAEHFYNGSSBGSS-KRWDZBQOSA-N
32681978
CC1=CC=C(C=C1)S(=O)(=O)N2CCCC2C3=CC=C(C=C3)F

Specifications

Ro 67-7476
298690-60-5
s-2-4-fluoro-phenyl-1-toluene-4-sulfonyl-pyrrolidine, pyrrolidine, 2-4-fluorophenyl-1-4-methylphenyl sulfonyl-, 2s, 2s-2-4-fluorophenyl-1-4-methylbenzenesulfonyl pyrrolidine, 2s-2-4-fluorophenyl-1-4-methylbenzene-1-sulfonyl pyrrolidine, 2s-2-4-fluorophenyl-1-4-methylphenyl sulfonyl-pyrrolidine
CC1=CC=C(C=C1)S(=O)(=O)N2CCCC2C3=CC=C(C=C3)F
319.394
319.39
50mg
C17H18FNO2S
DAEHFYNGSSBGSS-KRWDZBQOSA-N
(2S)-2-(4-fluorophenyl)-1-(4-methylphenyl)sulfonylpyrrolidine
32681978
>99%
Videos