missing translation for 'onlineSavingsMsg'
Learn More

Ro 64-5229, Tocris Bioscience™

Selective, non-competitive mGlu2 antagonist

Brand:  Tocris (Bio-Techne) 291350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462034

  • £855.54 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective, non-competitive mGlu2 antagonist. Inhibits GTPγ35S binding to mGlu2-containing membranes (IC50 = 0.11 μM).
Specifications

Chemical Identifiers

246852-46-0
352.259
d09ubs, 1-z-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1,2,4-triazole, 1-z-beta-cycloheptyloxy-2,6-dichlorostyryl-1h-1,2,4-triazole, z-1-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1h-1,2,4-triazole, 1-z-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole, 1-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole ro 64-5229
1-[(Z)-2-cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1,2,4-triazole
C17H19Cl2N3O
STCVFKBRXOEQRF-YBEGLDIGSA-N
10428048
C1CCCC(CC1)OC(=CN2C=NC=N2)C3=C(C=CC=C3Cl)Cl

Specifications

50mg
246852-46-0
d09ubs, 1-z-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1,2,4-triazole, 1-z-beta-cycloheptyloxy-2,6-dichlorostyryl-1h-1,2,4-triazole, z-1-2-cycloheptyloxy-2-2,6-dichlorophenyl ethenyl-1h-1,2,4-triazole, 1-z-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole, 1-2-cycloheptyloxy-2-2,6-dichloro-phenyl-vinyl-1h-1,2,4 triazole ro 64-5229
C1CCCC(CC1)OC(=CN2C=NC=N2)C3=C(C=CC=C3Cl)Cl
352.259
352.26
Ro 64-5229
C17H19Cl2N3O
STCVFKBRXOEQRF-YBEGLDIGSA-N
1-[(Z)-2-cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1,2,4-triazole
10428048
>99%
Videos
Documents