Learn More
Tocris Bioscience™ Ro 25-6981 maleate
NR2B-selective NMDA antagonist
Brand:  Tocris Bioscience™ 159450
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13459753
  • £570.72 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Potent and selective activity-dependent blocker of NMDA receptors containing the NR2B subunit. IC50 values are 0.009 and 52μM for cloned receptor subunit combinations NR1C/NR2B and NR1C/NR2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Specifications
Specifications
(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate
>98%
Observed MW: 455.55
Soluble to 100mM in DMSO
1312991-76-6
C22H29NO2.C4H4O4