Learn More
Tocris Bioscience™ Ro 25-6981 maleate
GluN2B-selective NMDA antagonist
Brand:  Tocris Bioscience™ 15941
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13439753
  • £79.65 / 1mg
Estimated Shipment
28-06-2019
Description
Description
Potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate
>98%
Observed MW: 455.55
Soluble to 100mM in DMSO
1312991-76-6
C22H29NO2.C4H4O4