missing translation for 'onlineSavingsMsg'
Learn More

Ro 10-5824 dihydrochloride, Tocris Bioscience™

Selective D4 receptor partial agonist

Brand:  Tocris (Bio-Techne) 232950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431044

  • £401.31 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective dopamine D4 receptor partial agonist (EC50 = 205 nM) that binds with high affinity (Ki = 5.2 nM). Displays 250-fold selectivity over D3 receptors and > 1000-fold selectivity over D2, D1 and D5 receptors.
Specifications

Chemical Identifiers

189744-94-3
353.291
ro 10-5824 dihydrochloride, 5-3,6-dihydro-4-phenyl-1 2h-pyridinyl methyl-2-methyl-4-pyrimidinamine dihydrochloride
2-methyl-5-[(4-phenyl-3,6-dihydro-2H-pyridin-1-yl)methyl]pyrimidin-4-amine;dihydrochloride
C17H22Cl2N4
UZHMRJRDYCRKIZ-UHFFFAOYSA-N
16759174
CC1=NC=C(C(=N1)N)CN2CCC(=CC2)C3=CC=CC=C3.Cl.Cl

Specifications

50mg
189744-94-3
ro 10-5824 dihydrochloride, 5-3,6-dihydro-4-phenyl-1 2h-pyridinyl methyl-2-methyl-4-pyrimidinamine dihydrochloride
CC1=NC=C(C(=N1)N)CN2CCC(=CC2)C3=CC=CC=C3.Cl.Cl
353.291
353.29
Ro 10-5824 dihydrochloride
C17H22Cl2N4
UZHMRJRDYCRKIZ-UHFFFAOYSA-N
2-methyl-5-[(4-phenyl-3,6-dihydro-2H-pyridin-1-yl)methyl]pyrimidin-4-amine;dihydrochloride
16759174
>99%
Videos
Documents