Learn More
Ro 04-6790, Tocris Bioscience™
Potent and selective 5-HT6 antagonist
Brand:  Tocris (Bio-Techne) 291150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13442034
  • £649.74 / 50mg
Estimated Shipment date
from Supplier 18-12-2019
Description
Description
Potent and selective 5-HT6 receptor antagonist (pKi values are 7.26 and 7.35 at rat and human 5-HT6 receptors respectively). Displays no affinity at a range of other receptors (IC50 > 10 μM). Induces stretching behavior in rats in vivo.
Specifications
Chemical Identifiers
202466-68-0
308.36
4-amino-n-2,6-bis methylamino-4-pyrimidinyl-benzenesulfonamide, 4-amino-n-2,6-bis methylamino pyrimidin-4-yl benzenesulfonamide, 4-amino-n-2,6-bis-methylamino-pyrimidin-4-yl-benzenesulfonamide, 4-amino-n-2,6-bis methylamino-4-pyrimidinyl benzenesulfonamide dihydrobromide, lopac-r-140, biomol-nt_000164, d0p8ja, gtpl274, 4-amino-n-2-methylamino-pyrimidin-4-yl-benzenesulfonamide
4-amino-N-[2,6-bis(methylamino)pyrimidin-4-yl]benzenesulfonamide
C12H16N6O2S
JELFWSXQTXRMAJ-UHFFFAOYSA-N
5312145
CNC1=CC(=NC(=N1)NC)NS(=O)(=O)C2=CC=C(C=C2)N
Specifications
50mg
202466-68-0
4-amino-n-2,6-bis methylamino-4-pyrimidinyl-benzenesulfonamide, 4-amino-n-2,6-bis methylamino pyrimidin-4-yl benzenesulfonamide, 4-amino-n-2,6-bis-methylamino-pyrimidin-4-yl-benzenesulfonamide, 4-amino-n-2,6-bis methylamino-4-pyrimidinyl benzenesulfonamide dihydrobromide, lopac-r-140, biomol-nt_000164, d0p8ja, gtpl274, 4-amino-n-2-methylamino-pyrimidin-4-yl-benzenesulfonamide
CNC1=CC(=NC(=N1)NC)NS(=O)(=O)C2=CC=C(C=C2)N
308.36
308.36
Ro 04-6790
C12H16N6O2S
JELFWSXQTXRMAJ-UHFFFAOYSA-N
4-amino-N-[2,6-bis(methylamino)pyrimidin-4-yl]benzenesulfonamide
5312145
>98%