missing translation for 'onlineSavingsMsg'
Learn More

Ro 04-5595 hydrochloride, Tocris Bioscience™

GluN2B-selective NMDA antagonist

Brand:  Tocris (Bio-Techne) 200510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13450384

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective antagonist for GluN2B (formally NR2B) containing NMDA receptors (Ki = 31 nM).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Specifications

Chemical Identifiers

64047-73-0
368.298
ro 04-5595 hydrochloride, 1-2-4-chlorophenyl ethyl-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-3,4-dihydro-1h-isoquinolin-7-ol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol hydrochloride
1-[2-(4-chlorophenyl)ethyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
C19H23Cl2NO2
NVIPBLQAFKRFSZ-UHFFFAOYSA-N
16759151
CN1CCC2=CC(=C(C=C2C1CCC3=CC=C(C=C3)Cl)O)OC.Cl

Specifications

10mg
64047-73-0
ro 04-5595 hydrochloride, 1-2-4-chlorophenyl ethyl-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-3,4-dihydro-1h-isoquinolin-7-ol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol hydrochloride
CN1CCC2=CC(=C(C=C2C1CCC3=CC=C(C=C3)Cl)O)OC.Cl
368.298
368.3
Ro 04-5595 hydrochloride
C19H23Cl2NO2
NVIPBLQAFKRFSZ-UHFFFAOYSA-N
1-[2-(4-chlorophenyl)ethyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
16759151
>98%
Videos
Documents