missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF213, Mouse anti-Human, Clone: 5C12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00057674-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LSPENAKLLSTFLNQTGLDAFLLELHEMIILKLKNPQTQTEERFRPQWSLRDTLVSYMQTKESEILPEMASQFPEEILLASCVSVWKTAAVLKWNRESpecifications
| RNF213 | |
| Monoclonal | |
| Unconjugated | |
| ring finger protein 213 | |
| C17orf27/KIAA1554 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 5C12 | |
| In 1x PBS, pH 7.4 | |
| NM_020914 | |
| RNF213 | |
| C17orf27 (NP_065965, 2016 a.a. ∼ 2112 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 57674 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction