missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ RNASET2 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human RNASET2 isoform 1 (Met 1- His 256) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 13509H08B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15286345

  • £398.71 / 25µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH: 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
8635
≥95% by SDS-PAGE
Human
<1 EU/μg
RNASET2
Other Proteins
RNASET2 Recombinant Human Protein
Lyophilized
28.5kd
5 x 50μg
Full Length, Recombinant
RNASET2 Recombinant Human Protein, His Tag
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.