missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ RNASET2 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human RNASET2 isoform 1 (Met 1- His 256) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 13509H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216355

  • £641.26 / 50µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH: 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
8635
≥95% by SDS-PAGE
Human
<1 EU/μg
RNASET2
Other Proteins
RNASET2 Recombinant Human Protein
Lyophilized
28.5kd
50μg
Full Length, Recombinant
RNASET2 Recombinant Human Protein, His Tag
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.