missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase I (10 U/μL)

Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Brand:  Thermo Scientific™ EN0601

80.07 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Code : 88

Additional Details : Weight : 1.51950kg

 View more versions of this product

Product Code. 10568930

  • £93.00 / 1000 units
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C
Specifications

Specifications

10 U/μL
RNase
1000 U
RNase I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.