Learn More
Thermo Scientific™ RNase AWAY™ Surface Decontaminant
Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.
Brand: Thermo Scientific™ 7002
40.32 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : 02
Additional Details : Weight : 0.54000kg
Description
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.
- Use on pipettes, gel boxes, or RNA or DNA prep areas
- Leaves no residue to interfere with gel polymerization or staining
- Chemically stable and nonabrasive
- Contains no strong acids
Specifications
11 | |
Spray Bottle | |
473 mL |
Sodium Hydroxide with Water | |
16 oz. |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide