Alfa Aesar™ RNA sample loading buffer


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J62832.S0

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00002kg


Quantity 2mL

Product Code. 15405139

Quantity Price
1 £ 51.52 / 2mL
Estimated Shipment date
from Supplier 07-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material RNA sample loading buffer
MDL Number MFCD00213041

Specification

Physical Form Liquid
Quantity 2mL
Health Hazard 3 GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H360-H351-H302-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Recommended Storage −30° to −10°C
Health Hazard 1 Danger
TSCA TSCA
MDL Number MFCD00213041
Chemical Name or Material RNA sample loading buffer
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement
H360-H351-H302-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

−30° to −10°C