missing translation for 'onlineSavingsMsg'
Learn More

RN 1747, Tocris Bioscience™

Selective TRPV4 agonist

Brand:  Tocris (Bio-Techne) 374550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463534

  • £479.72 / 50mg


Estimated Shipment
08-01-2019
Add to basket

Description

Description

Selective TRPV4 agonist (EC50 values are 0.77, 4.0 and 4.1μM for hTRPV4, mTRPV4 and rTRPV4 respectively). Displays selectivity over other TRP channels (EC50 values are 0.77, >30, >30 and >100μM for TRPV4, TRPM8, TRPV3 and TRPV1 receptors respectively). Also antagonizes TRPM8 at relevant concentrations (IC50 = 4μM).
Specifications

Chemical Identifiers

1024448-59-6
395.858
4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
C17H18ClN3O4S
ZNLVYSJQUMALEO-UHFFFAOYSA-N
5068295
C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]

Specifications

RN 1747
1024448-59-6
4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]
395.858
395.87
50mg
C17H18ClN3O4S
ZNLVYSJQUMALEO-UHFFFAOYSA-N
1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
5068295
>98%
Videos