missing translation for 'onlineSavingsMsg'
Learn More

RJR 2429 dihydrochloride, Tocris Bioscience™

Nicotinic receptor agonist

Brand:  Tocris (Bio-Techne) 127110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449033

  • £127.89 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent nAChR agonist that displays selectivity for α4β2 (Ki = 1 nM) and α1βγδ subtypes (EC50 values are 297 and 55 nM respectively). Induces dopamine release from striatal neurons (EC50 = 2 nM) and inhibits ion flux in thalamic neurons (IC50 = 154 nM). Also putative α3β4 agonist that potentiates catecholamine release.
Specifications

Chemical Identifiers

1021418-53-0
261.19
rjr 2429 dihydrochloride, +/--2-3-pyridinyl-1-azabicyclo 2.2.2 octane dihydrochloride
2-pyridin-3-yl-1-azabicyclo[2.2.2]octane;dihydrochloride
C12H18Cl2N2
PIUNXHHZRIUBOV-UHFFFAOYSA-N
56972204
C1CN2CCC1CC2C3=CN=CC=C3.Cl.Cl

Specifications

10mg
1021418-53-0
rjr 2429 dihydrochloride, +/--2-3-pyridinyl-1-azabicyclo 2.2.2 octane dihydrochloride
C1CN2CCC1CC2C3=CN=CC=C3.Cl.Cl
261.19
261.19
RJR 2429 dihydrochloride
C12H18Cl2N2
PIUNXHHZRIUBOV-UHFFFAOYSA-N
2-pyridin-3-yl-1-azabicyclo[2.2.2]octane;dihydrochloride
56972204
>98%
Videos
Documents