missing translation for 'onlineSavingsMsg'
Learn More

Rituximab Mouse, FITC, Monoclonal Antibody, AcroBiosystems

Mouse Monoclonal Antibody

£319.00 - £2160.00

Specifications

Antigen Rituximab
Classification Monoclonal
Conjugate FITC
Host Species Hybridoma
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17136651
View Documents
ACROBiosystems
RIB-FY35c-25ug
25 μg
£319.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17126651
View Documents
ACROBiosystems
RIB-FY35c-200ug
200 μg
£2160.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rituximab Monoclonal antibody specifically detects Rituximab.
Specifications

Specifications

Rituximab
FITC
RUO
Primary
Monoclonal
Hybridoma
IgG1
-20°C, aviod light
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.