missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ RIPA Buffer with Triton™ X-100 + 10% glycerol, 2X

Brand:  Alfa Aesar™ J60423.AK

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15403199

  • £64.98 / 250mL


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Specifications

250mL
Liquid
Cell Lysis Reagent
2X
Documentation

Technical Documents