missing translation for 'onlineSavingsMsg'
Learn More

Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, ACROS Organics™

Brand:  Acros Organics 349290010

£18.76 valid until 30/06/2017
Log in to get the promotional price



UNSPSC : 41104018

Code : 29

Additional Details : Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10451402

  • £46.90 / 1g


Estimated Shipment
16-06-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

Rink amide benzhydrylamine polymer resin
MFCD00677979

Specifications

crosslinking: 1% DVB
0.5 to 0.9mmol/g
Glass bottle
Off-white to pinkish-brown
MFCD00677979
Rink amide benzhydrylamine polymer resin
200 to 400 mesh
1g
Beads

Safety and Handling

Product Identifier
  • Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5 mmol/g

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray
  • P280-Wear protective gloves/protective clothing/eye protection/face protection
  • P302+P352-IF ON SKIN: Wash with plenty of water/
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents

Special Offers

Special Offers