Learn More
Thermo Scientific Acros Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, Acros Organics
Brand:  Thermo Scientific Acros 349290050
Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, Acros Organics
Code : 29
Additional Details : Weight : 0.05500kg
Product Code. 10285821