missing translation for 'onlineSavingsMsg'
Learn More

Rimcazole dihydrochloride, Tocris Bioscience™

σ2 antagonist. Also DAT inhibitor

Brand:  Tocris (Bio-Techne) 149750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459433

  • £578.52 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Antagonist at σ receptors (IC50 values are 1480 and 386 nM at σ1 and σ2 receptors respectively). Also binds to the dopamine transporter (IC50 = 57.6 nM) and inhibits dopamine uptake. Reduces the stimulatory effects of cocaine in vivo.
Specifications

Chemical Identifiers

75859-03-9
394.384
rimcazole dihydrochloride, rimcazole hydrochloride, bw 234u dihydrochloride, rimcazole hydrochloride usan, rimcazole hcl, 9-3-cis-3,5-dimethyl-1-piperazinyl propyl-9h-carbazole dihydrochloride, 9-3-cis-3,5-dimethyl-1-piperazinyl propyl carbazole dihydrochloride, cis-9-3-3,5-dimethyl-1-piperazinyl propyl-9h-carbazole dihydrochloride, dsstox_cid_25681, dsstox_rid_82704
9-[3-[(3R,5S)-3,5-dimethylpiperazin-1-yl]propyl]carbazole;dihydrochloride
C21H29Cl2N3
ZWHRZGHGYMUSRM-VWDRLOGHSA-N
53388
CC1CN(CC(N1)C)CCCN2C3=CC=CC=C3C4=CC=CC=C42.Cl.Cl

Specifications

Rimcazole dihydrochloride
75859-03-9
rimcazole dihydrochloride, rimcazole hydrochloride, bw 234u dihydrochloride, rimcazole hydrochloride usan, rimcazole hcl, 9-3-cis-3,5-dimethyl-1-piperazinyl propyl-9h-carbazole dihydrochloride, 9-3-cis-3,5-dimethyl-1-piperazinyl propyl carbazole dihydrochloride, cis-9-3-3,5-dimethyl-1-piperazinyl propyl-9h-carbazole dihydrochloride, dsstox_cid_25681, dsstox_rid_82704
CC1CN(CC(N1)C)CCCN2C3=CC=CC=C3C4=CC=CC=C42.Cl.Cl
394.384
394.39
50mg
C21H29Cl2N3
ZWHRZGHGYMUSRM-VWDRLOGHSA-N
9-[3-[(3R,5S)-3,5-dimethylpiperazin-1-yl]propyl]carbazole;dihydrochloride
53388
>98%
Videos