missing translation for 'onlineSavingsMsg'
Learn More

ALLSIGNS International™ Rigid Plastic Safety Sign

Brand:  ALLSIGNS International™ C09R

UNSPSC : 55121700

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12325538

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

400mm
Safety Sign
PVC (Polyvinyl Chloride)
400 x 300mm
Hazardous Area
300mm
1/Ea.
Documentation

Technical Documents