missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Rigid Plastic Safety Prohibition Sign

Brand:  Allsigns International™ EC45R

UNSPSC : 55121704

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12335758

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

Material: Rigid Plastic
Do Not Touch
300mm
200mm
Safety Prohibition Sign
Plastic
Documentation

Technical Documents