missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Rigid Photoluminescent Aluminum Right Warning Strips

Brand:  Brady™ 251944

Code : NEW

Additional Details : Weight : 0.06100kg

 View more versions of this product

Product Code. 11751036

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1m x 60mm
Red
Warning Strips
0.148mm
1Each
Rigid Photoluminescent Aluminum
Exit and directional signs
Self
Videos