missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rift Valley Fever Virus Nucleocapsid Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1611
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Rift Valley Fever Virus Nucleocapsid Synthetic Peptide, PEP-1611, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of Rift Valley Fever Virus.For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37ºC.Specifications
Virus | |
Rift Valley Fever Virus Nucleocapsid | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
0.02% sodium azide | |
50 μg | |
PBS |
purified | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
No | |
Peptide | |
RUO - research use only |