Learn More
Thermo Scientific™ Richard-Allan Scientific™ Fine Cutting Tool Kit
Improve consistency in grossing with the Thermo Scientific™ Richard-Allan Scientific™ Fine Cutting Tool Kit.
Brand: Thermo Scientific™ 9813
Additional Details : Weight : 0.01000kg
Description
Improve consistency in grossing with the Thermo Scientific™ Richard-Allan Scientific™ Fine Cutting Tool Kit.
- User may choose the correct tool to gross specimens at 3, 4 or 5mm thickness
- Easy to clean, chemical resistant, and autoclavable
Includes:Hex wrench and a ten-pack of high profile disposable blades.
Specifications
Fine Cutting Tool Kit, 3mm | |
Fine Cutting Tool Kit |