missing translation for 'onlineSavingsMsg'
Learn More

Ribonucleic acid from Baker's yeast, Thermo Scientific Chemicals

Catalog Number p-7042055
Click to view available options
Quantity:
100 mg
500 mg
1 g
63231-63-0
MFCD00132196
Ribonucleic acid
63231-63-0
MFCD00132196
Ribonucleic acid

Ribonucleic acid (RNA) from baker?s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ribonucleic acid (RNA) from baker′s yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.

Solubility
It is soluble in water.

Notes
Prepared by a modification of Crestfield, A.M., et al., J. Biol. Chem., 216, 185 (1955). Suitable for use as a ribonuclease substrate. Not suitable for use as a carrier in nucleic acid purification and precipitation
TRUSTED_SUSTAINABILITY
Color White to Brown
Quantity 100 mg
Solubility Information It is soluble in water.
Physical Form Powder
Chemical Name or Material Ribonucleic acid from Baker's yeast

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'