missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min., Thermo Scientific Chemicals

Catalog Number p-7057568
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6184-17-4
C17H24N4NaO11P
514.36
MFCD00150993
LQVGUKOCMOKKJU-FTNKLFHDNA-M
riboflavin 5'-monophosphate sodium salt dihydrate
131856174
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1
This item is not returnable. View return policy
6184-17-4
C17H24N4NaO11P
514.36
MFCD00150993
LQVGUKOCMOKKJU-FTNKLFHDNA-M
riboflavin 5'-monophosphate sodium salt dihydrate
131856174
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color Yellow to Orange
Quantity 10 g
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water at 50mg/mL
Formula Weight 514.36 (478.34 Anhydrous)
Physical Form Powder
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'